missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1,2,3,4-tetrahydroquinoline, 98%

Catalog No. AAA1524518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1524518 50 g
AAA1524509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1524518 Supplier Thermo Scientific Chemicals Supplier No. A1524518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 91-61-2 | C10H13N | 147.22 g/mol

6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 91-61-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023887
InChI Key XOKMRXSMOHCNIX-UHFFFAOYSA-N
Synonym civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa
PubChem CID 66678
IUPAC Name 6-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1=CC=C2NCCCC2=C1

Specifications

Melting Point 33°C to 37°C
Boiling Point 262°C to 263°C (712 mmHg)
Quantity 50 g
Beilstein 122515
Solubility Information Insoluble in water.
Formula Weight 147.22
Percent Purity 98%
Chemical Name or Material 6-Methyl-1,2,3,4-tetrahydroquinoline
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 202-083-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only