missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1,2,3,4-tetrahydroquinoline 97.0+%, TCI America™

Catalog No. M13025G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. M13025G missing translation for 'mfr' TCI America missing translation for 'supplierNo' M13025G

Chemical Identifiers

CAS 91-61-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023887
InChI Key XOKMRXSMOHCNIX-UHFFFAOYSA-N
Synonym civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa
PubChem CID 66678
IUPAC Name 6-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1=CC=C2NCCCC2=C1

Specifications

Melting Point 35°C
Color Yellow
Quantity 5 g
Formula Weight 147.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Methyl-1,2,3,4-tetrahydroquinoline
EINECSNumber (5)-0803
TSCA Yes