missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxyquinoline-4-carboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 280°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 203.20 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Methoxyquinoline-4-carboxylic Acid |
Safety and Handling
| TSCA | No |