missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyflavanone 98.0+%, TCI America™

Numéro de catalogue. M14035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M14035G 5 g
M14031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M14035G Fournisseur TCI America Code fournisseur M14035G
Il en reste null

Identifiants chimiques

CAS 3034-04-6
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
MDL Number MFCD00017484
InChI Key YURQMHCZHLMHIB-UHFFFAOYNA-N
Synonym 6-methoxyflavanone, 6-methoxy-2-phenylchroman-4-one, 6-methoxy-2-phenyl-chroman-4-one, 6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one, flavanone, 6-methoxy, acmc-1adv8, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one #, 2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-6-methoxy-2-phenyl
PubChem CID 97860
IUPAC Name 6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3

Spécifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 254.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxyflavanone