missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-3-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. M23475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23475G 5 g
M23471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23475G Supplier TCI America Supplier No. M23475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65873-72-5
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD02683446
InChI Key CTAIEPPAOULMFY-UHFFFAOYSA-N
Synonym 6-methoxynicotinaldehyde, 6-methoxy-3-pyridinecarboxaldehyde, 5-formyl-2-methoxypyridine, 2-methoxypyridine-5-carboxaldehyde, 5-formyl-2-methoxy-pyridine, 6-methoxy-pyridine-3-carbaldehyde, 6-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methoxy, 6-methoxy-3-nicotinaldehyde, pubchem5152
PubChem CID 3364576
IUPAC Name 6-methoxypyridine-3-carbaldehyde
SMILES COC1=CC=C(C=O)C=N1

Specifications

Melting Point 51°C
Color White
Quantity 5 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-3-pyridinecarboxaldehyde