missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthoic Acid 97.0+%, TCI America™

Catalog No. M14185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14185G 5 g
M141825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14185G Supplier TCI America Supplier No. M14185G

Chemical Identifiers

CAS 2471-70-7
Molecular Formula C12H9O3
Molecular Weight (g/mol) 201.20
MDL Number MFCD00092750
InChI Key YZBILXXOZFORFE-UHFFFAOYSA-M
Synonym 6-methoxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-methoxy, unii-65ih1sv66a, 6-methoxy-2-naphthoicacid, 6-methoxy-2-naphthalenecarboxylic acid, 6-methoxy-naphthalene-2-carboxylic acid, naproxen specified impurity o ep, naproxen impurity o, 2-methoxy-6-naphthalenecarboxylic acid, zlchem 1096
PubChem CID 349181
IUPAC Name 6-methoxynaphthalene-2-carboxylate
SMILES COC1=CC=C2C=C(C=CC2=C1)C([O-])=O

Specifications

Quantity 5 g
Formula Weight 202.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-2-naphthoic Acid