missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthoic acid Pharmaceutical Impurity Standard, ≥96.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013688
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013688 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013688 Supplier Merck Emd Millipore Supplier No. 0754750MG
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 2471-70-7
Linear Formula CH3 OC10H6 CO2 H
MDL Number MFCD00092750
Melting Point 201°C to 206°C
Synonym Naproxen impurity O (PhEur); 6-Methoxy-2-naphthalenecarboxylic acid
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 202.21
Percent Purity ≥96.5% (GC)