missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthaldehyde 97.0+%, TCI America™

Catalog No. M10905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10905G 5 g
M109025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10905G Supplier TCI America Supplier No. M10905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3453-33-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00081152
InChI Key VZBLASFLFFMMCM-UHFFFAOYSA-N
Synonym 6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395
PubChem CID 76991
IUPAC Name 6-methoxynaphthalene-2-carbaldehyde
SMILES COC1=CC=C2C=C(C=O)C=CC2=C1

Specifications

Melting Point 82°C
Boiling Point 196°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-2-naphthaldehyde