missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1,2,3,4-tetrahydroisoquinoline 96.0+%, TCI America™

Catalog No. M27811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27811G Supplier TCI America Supplier No. M27811G
Only null left

Specifications

CAS 42923-77-3
Color Yellow
Boiling Point 144°C
Molecular Formula C10H13NO
Quantity 1 g
MDL Number MFCD01717129
Synonym Longimammatine
InChI Key YYTAYINRPUJPNH-UHFFFAOYSA-N
SMILES COC1=CC=C2CNCCC2=C1
IUPAC Name 6-methoxy-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol) 163.22
PubChem CID 39356
Formula Weight 163.22
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroisoquinoline
Show More Show Less
RTECSNumber NX5082000
TSCA No