missing translation for 'onlineSavingsMsg'
Learn More

6-Iodoindazole 98.0+%, TCI America™

Catalog No. I09335G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09335G 5 g
I09331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09335G Supplier TCI America Supplier No. I09335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 261953-36-0
Molecular Formula C7H5IN2
Molecular Weight (g/mol) 244.035
MDL Number MFCD04114695
InChI Key RSGAXJZKQDNFEP-UHFFFAOYSA-N
Synonym 6-iodoindazole, 1h-indazole, 6-iodo, 6-iodo 1h indazole, 6-iodo-indazole, pubchem18081, indazole, 6-iodo
PubChem CID 12991241
IUPAC Name 6-iodo-1H-indazole
SMILES C1=CC2=C(C=C1I)NN=C2

Specifications

Melting Point 209°C
Quantity 5 g
Formula Weight 244.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Iodoindazole