missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Iodo-2-methyl-1,3-benzothiazole, 90%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 90% |
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 275.11 |
| Percent Purity | 90% |
| Chemical Name or Material | 6-Iodo-2-methyl-1, 3-benzothiazole |