missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-1,4-benzodioxane, 95%, remainder mainly 5-isomer

Catalog No. AAB2012714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2012714 25 g
AAB2012706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2012714 Supplier Thermo Scientific Chemicals Supplier No. B2012714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57744-67-9 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57744-67-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00221460
InChI Key HZTMYTXWFHBHDC-UHFFFAOYSA-N
Synonym 3,4-ethylenedioxyiodobenzene, 6-iodo-2,3-dihydrobenzo b 1,4 dioxine, 6-iodobenzodioxane, acmc-1azse, 6-iodo-1,4-benzodioxane, 6-iodo-2h,3h-benzo e 1,4-dioxin, 2,3-dihydro-6-iodobenzo 1,4 dioxin, 1,4-benzodioxin,2,3-dihydro-6-iodo
PubChem CID 2776176
IUPAC Name 6-iodo-2,3-dihydro-1,4-benzodioxine
SMILES IC1=CC=C2OCCOC2=C1

Specifications

Boiling Point 100°C to 102°C (0.25 mmHg)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity 95%
Assay remainder mainly 5-isomer
Chemical Name or Material 6-Iodo-1,4-benzodioxane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only