missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxypyridine-3-boronic acid pinacol ester, 97%

Catalog No. AAH51673MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51673MD 250 mg
AAH5167303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51673MD Supplier Thermo Scientific Chemicals Supplier No. H51673MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1054483-78-1 | C11H16BNO3 | 221.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1054483-78-1
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD06795671
InChI Key WAUWXCUPDOXYKS-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2 1h-one, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb145, pyridin-2-ol-5-boronic acid pinacol ester
PubChem CID 17750253
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyridin-2-one
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(O)C=C1

Specifications

Quantity 250 mg
Formula Weight 221.06
Percent Purity 97%
Chemical Name or Material 6-Hydroxypyridine-3-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only