missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxybenzothiazole 96.0+%, TCI America™

Numéro de catalogue. H13805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H13805G 5 g
H13801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H13805G Fournisseur TCI America Code fournisseur H13805G
Il en reste null

Identifiants chimiques

CAS 13599-84-3
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.18
MDL Number MFCD00057883
InChI Key ORIIXCOYEOIFSN-UHFFFAOYSA-N
Synonym 6-Benzothiazolol
PubChem CID 151811
IUPAC Name 1,3-benzothiazol-6-ol
SMILES OC1=CC=C2N=CSC2=C1

Spécifications

Melting Point 187°C
Color Yellow
Quantity 5 g
Formula Weight 151.18
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxybenzothiazole