missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxybenzothiazole 96.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 187°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 151.18 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Hydroxybenzothiazole |
Sécurité et manipulation
| TSCA | No |