missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-4-pyrimidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. H1392200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1392200MG 200 mg
H13921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1392200MG Supplier TCI America Supplier No. H1392200MG

Chemical Identifiers

CAS 6299-87-2
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00023267
InChI Key QYGHXDAYBIFGKI-UHFFFAOYSA-N
Synonym 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid
PubChem CID 239692
IUPAC Name 6-oxo-3,6-dihydropyrimidine-4-carboxylic acid
SMILES OC(=O)C1=CC(=O)N=CN1

Specifications

Melting Point 267°C
Quantity 200 mg
Formula Weight 140.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-4-pyrimidinecarboxylic Acid