missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxy-2-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Chemical Name or Material | 6-Hydroxy-2-naphthaleneboronic Acid (contains varying amounts of Anhydride) |
| CAS | 173194-95-1 |
| Melting Point | 224°C |
| Molecular Formula | C10H9BO3 |
| Quantity | 1 g |
| MDL Number | MFCD00092986 |
| Synonym | 6-hydroxy-2-naphthaleneboronic acid, 6-hydroxynaphthalen-2-yl boronic acid, 6-hydroxynaphthalene-2-boronic acid, 6-hydroxy-2-naphthyl boronic acid, 6-hydroxy-2-naphthyleneboronic acid, boronic acid,b-6-hydroxy-2-naphthalenyl, 6-hydroxy-2-naphtaleneboronic acid, 6-hydroxy-2-naphthalenyl boronic acid, boronic acid, 6-hydroxy-2-naphthalenyl, pubchem7886 |
| InChI Key | SNJANQMHRGSHFN-UHFFFAOYSA-N |
| SMILES | B(C1=CC2=C(C=C1)C=C(C=C2)O)(O)O |
| IUPAC Name | (6-hydroxynaphthalen-2-yl)boronic acid |
| Show More |