missing translation for 'onlineSavingsMsg'
Learn More

6-(Fmoc-amino)hexanoic acid, 98%

Catalog No. AAH6314814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6314814 25 g
AAH6314803 1 g
AAH6314806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6314814 Supplier Thermo Scientific Chemicals Supplier No. H6314814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88574-06-5 | C21H23NO4 | 353.418 g/mol

6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 88574-06-5
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.418
MDL Number MFCD00077045
InChI Key FPCPONSZWYDXRD-UHFFFAOYSA-N
Synonym fmoc-6-ahx-oh, fmoc-epsilon-acp-oh, n-fmoc-6-aminohexanoic acid, fmoc-, a-acp-oh, 6-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid, hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino
PubChem CID 2756087
IUPAC Name 6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCC(=O)O

Specifications

Color White
Linear Formula FmocNH(CH2)5CO2H
Quantity 25 g
Beilstein 5883220
Solubility Information Soluble in 1% acetic acid,water and 1mmol in 2mL DMF clearly soluble.
Formula Weight 353.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-(Fmoc-amino)hexanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only