missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoropyridine-3-boronic acid, 95%

Catalog No. AAL20387MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL20387MD 250 mg
AAL2038703 1 g
AAL2038706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL20387MD Supplier Thermo Scientific Chemicals Supplier No. L20387MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 351019-18-6 | C5H5BFNO2 | 140.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351019-18-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
MDL Number MFCD03411559
InChI Key OJBYZWHAPXIJID-UHFFFAOYSA-N
Synonym 2-fluoropyridine-5-boronic acid, 2-fluoro-5-pyridylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridine-3-boronic acid, 2-fluoro-5-pyridineboronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridin-5-boronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoro-3-pyridineboronic acid
PubChem CID 2783397
IUPAC Name (6-fluoropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=C(F)N=C1

Specifications

Melting Point 177°C to 178°C (decomposition)
Quantity 250 mg
Beilstein 8974962
Formula Weight 140.91
Percent Purity 95%
Chemical Name or Material 6-Fluoropyridine-3-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 627-412-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only