missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorochroman-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. F10865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10865G 5 g
F108625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10865G Supplier TCI America Supplier No. F10865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99199-60-7
Molecular Formula C10H9FO3
Molecular Weight (g/mol) 196.177
MDL Number MFCD07778357
InChI Key ZNJANLXCXMVFFI-UHFFFAOYSA-N
Synonym 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic Acid
PubChem CID 15382843
IUPAC Name 6-fluoro-3,4-dihydro-2H-chromene-2-carboxylic acid
SMILES C1CC2=C(C=CC(=C2)F)OC1C(=O)O

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluorochroman-2-carboxylic Acid