missing translation for 'onlineSavingsMsg'
Learn More

6-(Dimethylamino)fulvene 98.0+%, TCI America™

Catalog No. D27855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27855G 5 g
D27851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27855G Supplier TCI America Supplier No. D27855G

Chemical Identifiers

CAS 696-68-4
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00013275
InChI Key UJXXVGXQTXQALL-UHFFFAOYSA-N
Synonym 5-(Dimethylaminomethylene)-1,3-cyclopentadiene
PubChem CID 136523
IUPAC Name 1-cyclopenta-2,4-dien-1-ylidene-N,N-dimethylmethanamine
SMILES CN(C)C=C1C=CC=C1

Specifications

Melting Point 67°C
Quantity 5 g
Formula Weight 121.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Dimethylamino)fulvene
TSCA No
Recommended Storage Freezer