missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-3-methanol, 98%

Catalog No. AAL1928514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1928514 25 g
AAL1928506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1928514 Supplier Thermo Scientific Chemicals Supplier No. L1928514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21543-49-7 | C6H6ClNO | 143.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21543-49-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD01075226
InChI Key GOXYBEXWMJZLJB-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol
PubChem CID 177161
IUPAC Name (6-chloropyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)Cl

Specifications

Melting Point 46°C to 49°C
Boiling Point 120°C (0.4 mmHg)
Quantity 25 g
Beilstein 1525126
Formula Weight 143.57
Percent Purity 98%
Chemical Name or Material 6-Chloropyridine-3-methanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only