missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyridine-3-carboxaldehyde, 97%

Catalog No. AC398100050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398100050 5 g Glass bottle
AC398100010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398100050 Supplier Thermo Scientific Chemicals Supplier No. 398100050

CAS: 23100-12-1 | C6H4ClNO | 141.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23100-12-1
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.56
MDL Number MFCD03095223
InChI Key AFWWKZCPPRPDQK-UHFFFAOYSA-N
Synonym 6-chloronicotinaldehyde, 2-chloropyridine-5-carbaldehyde, 2-chloropyridine-5-carboxaldehyde, 6-chloropyridine-3-carboxaldehyde, 2-chloro-5-formylpyridine, 6-chloro-pyridine-3-carbaldehyde, 6-chloro-3-pyridinecarboxaldehyde, 2-chloro-5-pyridine carbaldehyde, 6-chloro-pyridin-3-carbaldehyde, 2-chloro-5-pyridinecarboxaldehyde
PubChem CID 2764053
IUPAC Name 6-chloropyridine-3-carbaldehyde
SMILES C1=CC(=NC=C1C=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77°C to 81°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 141.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-chloropyridine-3-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only