missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine riboside, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6461218
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6461218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6461218 Supplier Thermo Scientific Chemicals Supplier No. J6461218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2004-06-0 | Molecular Formula: C10H11ClN4O4 | Molecular Weight: 286.672 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2004-06-0
Molecular Formula C10H11ClN4O4
Molecular Weight (g/mol) 286.672
MDL Number MFCD00005738
InChI Key XHRJGHCQQPETRH-VKZRYZQFSA-N
Synonym 6-chloropurine ribonucleoside
PubChem CID 92022056
IUPAC Name (3S,4R,5S)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O

Specifications

Melting Point 158°C to 162°C
Color Yellow to Green
Quantity 50 g
Beilstein 40573
Formula Weight 286.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Chloropurine riboside
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-904-8
RTECSNumber UO7520800
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only