missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine, 99%

Catalog No. AAA1120214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1120214 25 g
AAA1120203 1 g
AAA1120206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1120214 Supplier Thermo Scientific Chemicals Supplier No. A1120214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-42-3 | C5H3ClN4 | 154.557 g/mol

6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.

Solubility
Soluble in hot water and dimethyl formamide.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 87-42-3
Molecular Formula C5H3ClN4
Molecular Weight (g/mol) 154.557
MDL Number MFCD00075825
InChI Key ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Synonym 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
PubChem CID 5359277
IUPAC Name 6-chloro-7H-purine
SMILES C1=NC2=C(N1)C(=NC=N2)Cl

Specifications

Melting Point >300°C (decomposition)
Quantity 25 g
Beilstein 5774
Merck Index 14,2161
Solubility Information Soluble in hot water and dimethyl formamide.
Formula Weight 154.56
Percent Purity 99%
Chemical Name or Material 6-Chloropurine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-745-6
RTECSNumber UO7520000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only