missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinamide 98.0+%, TCI America™

Catalog No. C17255G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17255G Supplier TCI America Supplier No. C17255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6271-78-9
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.57
MDL Number MFCD00006242
InChI Key ZIJAZUBWHAZHPL-UHFFFAOYSA-N
Synonym 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
PubChem CID 80456
IUPAC Name 6-chloropyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(Cl)N=C1

Specifications

Color White
Quantity 5 g
Formula Weight 156.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloronicotinamide