missing translation for 'onlineSavingsMsg'
Learn More

6-chloronicotinamide, 98%, Thermo Scientific™

Catalog No. AC109560010
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109560010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC109560010 Supplier Thermo Scientific Chemicals Supplier No. 109560010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6271-78-9
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.57
MDL Number MFCD00006242
InChI Key ZIJAZUBWHAZHPL-UHFFFAOYSA-N
Synonym 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
PubChem CID 80456
IUPAC Name 6-chloropyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(Cl)N=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Cl(C5H3N)CONH2
Quantity 1g
Beilstein 22,44
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 6-Chloronicotinamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 228-456-8

RUO – Research Use Only