missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorométhyluracil 98,0+%, TCI America™

Numéro de catalogue. p-7124765
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C10575G 5 g
C105725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C10575G Fournisseur TCI America Code fournisseur C10575G

Identifiants chimiques

CAS 18592-13-7
Molecular Formula C5H5ClN2O2
Molecular Weight (g/mol) 160.56
MDL Number MFCD00010095
InChI Key VCFXBAPEXBTNEA-UHFFFAOYSA-N
Synonym 6-chloromethyl uracil, 6-chloromethyl pyrimidine-2,4 1h,3h-dione, 6-chloromethyluracil, 2,4 1h,3h-pyrimidinedione, 6-chloromethyl, 6-chloromethyl-1h-pyrimidine-2,4-dione, 6-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 6-chloromethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 4-chloromethyl uracil, 6-chloromethyl-uracil, 6-chloromethyl uracil?
PubChem CID 73273
IUPAC Name 6-(chlorométhyl)-1,2,3,4-tétrahydropyrimidine-2,4-dione
SMILES ClCC1=CC(=O)NC(=O)N1

Spécifications

Color Blanc-jaune
Quantity 5 g
Formula Weight 160.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material 6-Chloromethyluracil