missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%

Catalog No. AAH35567MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35567MD 250 mg
AAH3556703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH35567MD Supplier Thermo Scientific Chemicals Supplier No. H35567MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 138642-97-4 | C8H5ClN2O2 | 196.59 g/mol

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 138642-97-4
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD07021509
InChI Key LXRDUEDQFSBROM-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-chloro
PubChem CID 16413639
IUPAC Name 6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC(Cl)=CN12

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 196.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only