missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-methylpyridine-2-carboxylic acid, 95%

Catalog No. AAH6358406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6358406 5 g
AAH6358403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6358406 Supplier Thermo Scientific Chemicals Supplier No. H6358406

CAS: 324028-95-7 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 324028-95-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD09834827
InChI Key IDIRHQNTRMGBPJ-UHFFFAOYSA-N
Synonym 6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid
PubChem CID 23002381
IUPAC Name 6-chloro-4-methylpyridine-2-carboxylic acid
SMILES CC1=CC(=NC(=C1)Cl)C(=O)O

Specifications

Color White
Quantity 5 g
UN Number UN2811
Formula Weight 171.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Chloro-4-methylpyridine-2-carboxylic acid
Hazard Category H301-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only