missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-(2-Chlorophenyl)Quinazoline-2-carboxaldehyde, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022894
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022894 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022894 Supplier Merck Emd Millipore Supplier No. BP376

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 93955-15-8
Molecular Formula C15H8Cl2N2O
Linear Formula C15H8Cl2N2O
MDL Number MFCD01570906
Quantity 50 mg
Synonym 6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxaldehyde
Formula Weight 303.14
Grade BP