missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-pyridinemethanol 98.0+%, TCI America™

Catalog No. C18975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18975G 5 g
C189725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18975G Supplier TCI America Supplier No. C18975G

Chemical Identifiers

CAS 21543-49-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
InChI Key GOXYBEXWMJZLJB-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol
PubChem CID 177161
IUPAC Name (6-chloropyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)Cl

Specifications

Melting Point 54°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-3-pyridinemethanol