missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-iodo-3-hydroxypyridine 98.0+%, TCI America™

Catalog No. C33665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33665G 5 g
C33661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33665G Supplier TCI America Supplier No. C33665G

Chemical Identifiers

CAS 188057-26-3
Molecular Formula C5H3ClINO
Molecular Weight (g/mol) 255.439
MDL Number MFCD07636749
InChI Key FWIMPBYPMQSSCD-UHFFFAOYSA-N
PubChem CID 23433158
IUPAC Name 6-chloro-2-iodopyridin-3-ol
SMILES C1=CC(=NC(=C1O)I)Cl

Specifications

Melting Point 193°C
Color Yellow
Quantity 5 g
Synonym 6-chloro-2-iodo-3-hydroxypyridine, 6-chloro-2-iodo-3-pyridinol, 6-chloro-2-iodo-pyridin-3-ol, 3-pyridinol, 6-chloro-2-iodo, 2-iodo-6-chloropyridine-3-ol, 3-pyridinol,6-chloro-2-iodo
Formula Weight 255.44
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2-iodo-3-hydroxypyridine