missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-hydroxypyridine 98.0+%, TCI America™

Catalog No. C107925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C107925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C107925G Supplier TCI America Supplier No. C107925G

Chemical Identifiers

CAS 16879-02-0
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006278
InChI Key CLNNBQDAAGDAHI-UHFFFAOYSA-N
Synonym 6-chloropyridin-2-ol, 2-chloro-6-hydroxypyridine, 6-chloro-2-pyridinol, 6-chloro-2-hydroxypyridine, 6-chloropyridin-2 1h-one, 2 1h-pyridinone, 6-chloro, 6-chloro-2 1h-pyridinone, 6-chloro-2-pyridone, 6-chloropyridn-2-ol, 2 1h-pyridone, 6-chloro
PubChem CID 85621
IUPAC Name 6-chloro-1H-pyridin-2-one
SMILES C1=CC(=O)NC(=C1)Cl

Specifications

Melting Point 130°C
Quantity 25 g
Formula Weight 129.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2-hydroxypyridine