missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals

Catalog No. AC297300250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297300250 25 g Glass bottle
AC297300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297300250 Supplier Thermo Scientific Chemicals Supplier No. 297300250

missing translation for 'validMainframeMsg'

CAS: 22876-20-6 | Molecular Formula: C7H4ClNOS | Molecular Weight: 185.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22876-20-6
Molecular Formula C7H4ClNOS
Molecular Weight (g/mol) 185.63
MDL Number MFCD01456482
InChI Key HAASPZUBSZGCKU-UHFFFAOYSA-N
Synonym 6-chloro-1,3-benzoxazole-2-thiol, 6-chloro-2-mercaptobenzoxazole, 6-chloro-2-benzoxazolethiol, 6-chlorobenzo d oxazole-2-thiol, 2-mercapto-6-chlorobenzoxazole, 6-chlorobenzoxazole-2 3h-thione, 6-chlorobenzo d oxazole-2 3h-thione, 6-chloro-benzooxazole-2-thiol, pubchem8714, 6-chlorobenzoxazole-2-thiol
PubChem CID 2119214
IUPAC Name 6-chloro-3H-1,3-benzoxazole-2-thione
SMILES C1=CC2=C(C=C1Cl)OC(=S)N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 229.0°C to 232.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in solutions of bases,soluble in ethanol
Formula Weight 185.63
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 6-Chloro-2-benzoxazolethiol, 99%
Show More Show Less
EINECSNumber 245-282-8

RUO – Research Use Only