missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,4-dimethoxypyrimidine 98.0+%, TCI America™

Catalog No. D13855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13855G 5 g
D13851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13855G Supplier TCI America Supplier No. D13855G

Chemical Identifiers

CAS 6320-15-6
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.58
MDL Number MFCD00006068
InChI Key JHNRTJRDRWKAIW-UHFFFAOYSA-N
Synonym 6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine
PubChem CID 80600
IUPAC Name 4-chloro-2,6-dimethoxypyrimidine
SMILES COC1=CC(Cl)=NC(OC)=N1

Specifications

Melting Point 74°C
Color White
Quantity 5 g
Formula Weight 174.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2,4-dimethoxypyrimidine