missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,4-dimethoxypyrimidine, 98+%

Catalog No. AAB2169614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169614 25 g
AAB2169606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2169614 Supplier Thermo Scientific Chemicals Supplier No. B2169614

CAS: 6320-15-6 | C6H7ClN2O2 | 174.58 g/mol

It is used in the synthesis of silyl and stannyl pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of silyl and stannyl pyrimidines.

Solubility
Soluble in chloroform, ethyl acetate and methanol.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6320-15-6
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.58
MDL Number MFCD00006068
InChI Key JHNRTJRDRWKAIW-UHFFFAOYSA-N
Synonym 6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine
PubChem CID 80600
IUPAC Name 4-chloro-2,6-dimethoxypyrimidine
SMILES COC1=CC(Cl)=NC(OC)=N1

Specifications

Melting Point 71°C to 73°C
Quantity 25 g
Beilstein 137581
Sensitivity Hygroscopic
Solubility Information Soluble in chloroform,ethyl acetate and methanol.
Formula Weight 174.59
Percent Purity ≥98%
Chemical Name or Material 6-Chloro-2,4-dimethoxypyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-669-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only