missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-1,3-benzothiazol-2-amine, 97%, Thermo Scientific™

Catalog No. btb04800da
Change view
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité Emballage
BTB04800DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB04800DA Supplier Thermo Scientific Chemicals Supplier No. BTB04800DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-24-9
Formule moléculaire C7H5ClN2S
Poids moléculaire (g/mol) 184.641
Numéro MDL MFCD00053557
Clé InChI VMNXKIDUTPOHPO-UHFFFAOYSA-N
Synonyme 2-amino-6-chlorobenzothiazole, 6-chlorobenzothiazol-2-ylamine, 2-benzothiazolamine, 6-chloro, 6-chlorobenzo d thiazol-2-amine, 6-chloro-benzothiazol-2-ylamine, unii-2u337t5ufg, 6-chloro-2-benzothiazolamine, benzothiazole, 2-amino-6-chloro, 6-chlorobenzothiazole-2-ylamine, c7h4brcln2s
PubChem CID 7226
Nom de l’IUPAC 6-chloro-1,3-benzothiazol-2-amine
SOURIRES C1=CC2=C(C=C1Cl)SC(=N2)N

Specifications

Nom ou substance chimique 6-Chloro-1, 3-benzothiazol-2-amine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 184.65
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.