missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoquinoline-5-carbonitrile, 95%

Catalog No. p-7039958
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59484MD 250 mg
AAH5948403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59484MD Supplier Thermo Scientific Chemicals Supplier No. H59484MD
Only null left

CAS: 1188365-70-9 | C10H5BrN2 | 233.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1188365-70-9
Molecular Formula C10H5BrN2
Molecular Weight (g/mol) 233.068
MDL Number MFCD22683072
InChI Key DTWHKRFZYKUTHB-UHFFFAOYSA-N
Synonym 6-Bromo-5-cyanoquinoline
PubChem CID 70817991
IUPAC Name 6-bromoquinoline-5-carbonitrile
SMILES C1=CC2=C(C=CC(=C2C#N)Br)N=C1

Specifications

Quantity 250 mg
UN Number UN3439
Beilstein 20114547
Formula Weight 233.07
Percent Purity 95%
Chemical Name or Material 6-Bromoquinoline-5-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only