missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoimidazo[1,2-a]pyrimidine 98.0+%, TCI America™

Catalog No. B55995G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55995G 5 g
B55991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55995G Supplier TCI America Supplier No. B55995G
Only null left

Chemical Identifiers

CAS 865156-68-9
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.023
MDL Number MFCD09261434
InChI Key BQMWMOQCMFLRQQ-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyrimidine, 6-bromo-imidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyrimidine, pubchem20931, acmc-209qa4, 6-bromo-imidazo 1,2-a pyr..., 6-bromanylimidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine,6-bromo
PubChem CID 26967622
IUPAC Name 6-bromoimidazo[1,2-a]pyrimidine
SMILES C1=CN2C=C(C=NC2=N1)Br

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyrimidine