missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanoyl Chloride 97.0+%, TCI America™

Catalog No. B204925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B204925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B204925G Supplier TCI America Supplier No. B204925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22809-37-6
Molecular Formula C6H10BrClO
Molecular Weight (g/mol) 213.499
MDL Number MFCD00000761
InChI Key HBPVGJGBRWIVSX-UHFFFAOYSA-N
Synonym hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride
PubChem CID 89848
IUPAC Name 6-bromohexanoyl chloride
SMILES C(CCC(=O)Cl)CCBr

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 3265
Formula Weight 213.50
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6-Bromohexanoyl Chloride