missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-chloroquinoline 98,0+%, TCI America™

Numéro de catalogue. B51665G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B51665G 5 g
B51661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B51665G Fournisseur TCI America Code fournisseur B51665G

Identifiants chimiques

CAS 65340-70-7
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.5
MDL Number MFCD00511001
InChI Key KJILYZMXTLCPDQ-UHFFFAOYSA-N
Synonym 4-chloro-6-bromoquinoline, 6-bromo-4-chloro-quinoline, quinoline, 6-bromo-4-chloro, pubchem20825, acmc-1bdfp, 4-choro-6-bromoquinoline, 6-bromo-4-chloroquinoline, 6-bromo-4-chloro quinoline, ksc493o7p
PubChem CID 5139537
IUPAC Name 6-bromo-4-chloroquinoline
SMILES C1=CC2=NC=CC(=C2C=C1Br)Cl

Spécifications

Melting Point 113°C
Quantity 5 g
Formula Weight 242.50
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 6-Bromo-4-chloroquinoline