missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2H-1,4-benzoxazin-3(4H)-one, 95%

Catalog No. AAH2749309
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2749309 10 g
AAH2749303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2749309 Supplier Thermo Scientific Chemicals Supplier No. H2749309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24036-52-0 | C8H6BrNO2 | 228.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24036-52-0
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.045
MDL Number MFCD00461173
InChI Key UQCFMEFQBSYDHY-UHFFFAOYSA-N
Synonym 6-bromo-2h-1,4-benzoxazin-3 4h-one, 6-bromo-2h-benzo b 1,4 oxazin-3 4h-one, 6-bromo-3-oxo-4h-benzo 1,4 oxazine, 6-bromo-1,4-benzoxazin-3-one, 6-bromo-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo, 6-bromo-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-4h-benzo 1,4 oxazin-3-one, pubchem16746
PubChem CID 16218142
IUPAC Name 6-bromo-4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=C(O1)C=CC(=C2)Br

Specifications

Melting Point 220°C to 225°C
Quantity 10 g
Formula Weight 228.04
Percent Purity 95%
Chemical Name or Material 6-Bromo-2H-1,4-benzoxazin-3(4H)-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only