missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. B37165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37165G 5 g
B37161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37165G Supplier TCI America Supplier No. B37165G

Chemical Identifiers

CAS 34160-40-2
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD02683546
InChI Key QWFHFNGMCPMOCD-UHFFFAOYSA-N
Synonym 6-bromo-2-pyridinecarboxaldehyde, 6-bromopicolinaldehyde, 6-bromo-2-formylpyridine, 6-bromopyridine-2-carboxaldehyde, 2-bromo-6-formylpyridine, 6-bromo-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-bromo, 2-bromopyridine-6-carboxaldehyde, 6-bromo-2-pyridine carboxaldehyde, 6-bromopicolylaldehyde
PubChem CID 2757009
IUPAC Name 6-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Br)C=O

Specifications

Melting Point 81°C
Quantity 5 g
Formula Weight 186.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-pyridinecarboxaldehyde
TSCA No
Recommended Storage Refrigerator