missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chlorobenzothiazole 96.0+%, TCI America™

Catalog No. B44485G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44485G 5 g
B44481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44485G Supplier TCI America Supplier No. B44485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80945-86-4
Molecular Formula C7H3BrClNS
Molecular Weight (g/mol) 248.522
MDL Number MFCD04971822
InChI Key IJQSMNIZBBEBKI-UHFFFAOYSA-N
Synonym 6-bromo-2-chlorobenzothiazole, 6-bromo-2-chlorobenzo d thiazole, 6-bromo-2-chloro-benzothiazole, 2-chloro-6-bromobenzothiazole, benzothiazole, 6-bromo-2-chloro, 6-bromo-2-chlorbenzothiazole, benzothiazole,6-bromo-2-chloro, zlchem 563, pubchem11132, acmc-1bkg3
PubChem CID 2049871
IUPAC Name 6-bromo-2-chloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC(=N2)Cl

Specifications

Melting Point 100°C
Color Yellow
Boiling Point 157°C
Quantity 5 g
Formula Weight 248.52
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-chlorobenzothiazole