missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%

Catalog No. AAH27460MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27460MD 250 mg
AAH2746003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27460MD Supplier Thermo Scientific Chemicals Supplier No. H27460MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072944-23-0 | C7H9BBrNO2 | 229.868 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072944-23-0
Molecular Formula C7H9BBrNO2
Molecular Weight (g/mol) 229.868
MDL Number MFCD09037491
InChI Key GXXITYQLWOMIEM-UHFFFAOYSA-N
Synonym 6-bromo-2,4-dimethylpyridine-3-boronic acid, 6-bromo-2,4-dimethylpyridin-3-yl boronic acid, acmc-2098oh, 6-bromo-2,4-dimethylpyridin-3-yl boronicacid
PubChem CID 44755186
IUPAC Name (6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1C)Br)C)(O)O

Specifications

Melting Point ∽145°C (decomposition)
Quantity 250 mg
Formula Weight 229.87
Percent Purity 95%
Chemical Name or Material 6-Bromo-2,4-dimethylpyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only