missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-tetralone, 96%, Thermo Scientific Chemicals

Catalog No. AAH5238103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5238103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5238103 Supplier Thermo Scientific Chemicals Supplier No. H5238103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66361-67-9 | Molecular Formula: C10H9BrO | Molecular Weight: 225.09 g/mol

6-bromo-1-tetralone used in Wittig-Horner reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-bromo-1-tetralone used in Wittig-Horner reaction.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.

Chemical Identifiers

CAS 66361-67-9
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.09
MDL Number MFCD04114378
InChI Key OSDHOOBPMBLALZ-UHFFFAOYSA-N
Synonym 6-bromo-1-tetralone, 6-bromo-3,4-dihydronaphthalen-1 2h-one, 6-bromotetral-1-one, 6-bromo-tetral-1-on, 6-bromotetralone, 1 2h-naphthalenone, 6-bromo-3,4-dihydro, 6-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 6-bromo-3,4-dihydro-1 2h-naphthalenone, 6-bromoteralone, 6-bromo-tetralone
PubChem CID 10105069
IUPAC Name 6-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES BrC1=CC2=C(C=C1)C(=O)CCC2

Specifications

Melting Point 40°C to 42°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 225.09
Percent Purity 96%
Chemical Name or Material 6-Bromo-1-tetralone
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only