missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 96%, Thermo Scientific Chemicals

Catalog No. AAH66700MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66700MD 250 mg
AAH6670003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66700MD Supplier Thermo Scientific Chemicals Supplier No. H66700MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 356560-80-0 | C6H4BrN3 | 198.023 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 356560-80-0
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.023
MDL Number MFCD08689534
InChI Key CXRXKDSDRWLKTK-UHFFFAOYSA-N
Synonym 6-bromo-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine, 6-bromo, 6-bromo-1,2,4-triazolo-1,5-a-pyridine, 6-bromo-1,2,4-triazolo 1,5-a pyridine, 1,2,4 triazolo 1,5-a pyridine,6-bromo, 6-bromanyl-1,2,4 triazolo 1,5-a pyridine, 6-bromo 1,2,4 triazolo 1,5-alpha pyridine, tert-butyl 6e-7-4-4-fluorophenyl-6-isopropyl-2-methyl methylsulfonyl amino pyrimidin-5-yl-3r,5s-3,5-dihydroxyhept-6-enoate
PubChem CID 11513934
IUPAC Name 6-bromo-[1,2,4]triazolo[1,5-a]pyridine
SMILES C1=CC2=NC=NN2C=C1Br

Specifications

Quantity 250 mg
Beilstein 10061274
Formula Weight 198.02
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 6-Bromo-[1,2,4]triazolo[1,5-a]pyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only