missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyladenine, 99%

Catalog No. AAA1467814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1467814 25 g
AAA1467803 1 g
AAA1467806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1467814 Supplier Thermo Scientific Chemicals Supplier No. A1467814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1214-39-7 | C12H11N5 | 225.26 g/mol

Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.

Notes
Incompatible with strong oxidizing agent.

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

Melting Point 229°C to 233°C
Assay Percent Range 99%
Quantity 25 g
Beilstein 19406
Solubility Information Soluble in water,methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Formula Weight 225.26
Percent Purity 99%
Chemical Name or Material 6-Benzyladenine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-927-5
RTECSNumber AU6252200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only