missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-3-chloro-1H-indazole, 97%

Catalog No. AAH6144506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6144506 5 g
AAH6144503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6144506 Supplier Thermo Scientific Chemicals Supplier No. H6144506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21413-23-0 | C7H6ClN3 | 167.60 g/mol

6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Sparingly soluble in water (0.62 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 21413-23-0
Molecular Formula C7H6ClN3
Molecular Weight (g/mol) 167.60
MDL Number MFCD07781648
InChI Key DRRARKIFTNKQDW-UHFFFAOYSA-N
Synonym 3-chloro-1h-indazol-6-amine, 6-amino-3-chloro-1h-indazole, 1h-indazol-6-amine,3-chloro, 6-amino-3-chloroindazole, 3-chloro-1h-indazole-6-ylamine, 3-chloro-1h-indazol-6-ylamine, 6-amino-3-chloro 1h indazole, pubchem23042, 3-chloro-indazol-6-amine
PubChem CID 14790453
IUPAC Name 3-chloro-2H-indazol-6-amine
SMILES NC1=CC2=NNC(Cl)=C2C=C1

Specifications

Quantity 5 g
Solubility Information Sparingly soluble in water (0.62g/L at 25°C).
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 6-Amino-3-chloro-1H-indazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only